Learn More
Microsolv Technology Corporation Caps Crimp Silver Red Rubber
Catalog No. 502755
Encompass_Preferred
Caps, Crimp Top, with Red Rubber / PTFE Septa, fitted in aluminum, Silver Caps. For use with 11mm Crimp or Snap Top, 12x32mm autosampler vials. BASIK Brand. 1000/CS.
Catalog No. 50-275-5 Supplier Microsolv Technology Corporation Supplier No. 9502C40TSM
Caps, Crimp Top, with Red Rubber / PTFE Septa, fitted in aluminum, Silver Caps. For use with 11mm Crimp or Snap Top, 12x32mm autosampler vials. BASIK Brand. 1000/CS.