Learn More
Microsolv Technology Corporation Caps, Crimp Top, with Red Rubber / PTFE Septa, fitted in aluminum, Gold Caps. For use with 11mm Crimp or Snap Top, 12x32mm autosampler vials. 1000/CS.
Catalog No. 502270366
Encompass_Preferred
The Rubber is softer than Natural Red Rubber and provides better sealing after injection. Great for GC.
This item is not returnable. View return policy
Catalog No. 50-227-0366 Supplier Microsolv Technology Corporation Supplier No. 9502C40TDM
The Rubber is softer than Natural Red Rubber and provides better sealing after injection. Great for GC.