Learn More
Microsolv Technology Corporation Caps, Crimp Top, with Red Rubber / PTFE Septa, fitted in aluminum, Red Caps. For use with 11mm Crimp or Snap Top, 12x32mm autosampler vials. 1000/CS.
Supplier:  Microsolv Technology Corporation 9502C40TRM
Encompass_Preferred
The Rubber is softer than Natural Red Rubber and provides better sealing after injection. Great for GC.
Catalog No. 50-227-0369