Learn More
Microsolv Technology Corporation Caps, Crimp Top, with Red Rubber / PTFE Septa, fitted in aluminum, Green Caps. For use with 11mm Crimp or Snap Top, 12x32mm autosampler vials. 100/PK.
Catalog No. 502270367
Encompass_Preferred
The Rubber is softer than Natural Red Rubber and provides better sealing after injection. Great for GC.
This item is not returnable. View return policy
Catalog No. 50-227-0367 Supplier Microsolv Technology Corporation Supplier No. 9502C40TG
The Rubber is softer than Natural Red Rubber and provides better sealing after injection. Great for GC.