Learn More
Microsolv Technology Corporation Caps, Crimp Top, with Red Rubber / PTFE Septa, fitted in aluminum, Sliver Caps. For use with 11mm Crimp or Snap Top, 12x32mm autosampler vials. 100/PK.
Catalog No. 502270370
Encompass_Preferred
Our Crimp Cap. The Rubber is softer than Natural Red Rubber and provides better sealing after injection. Great for GC. This is our most economical aluminum crimp cap.
This item is not returnable. View return policy
Catalog No. 50-227-0370 Supplier Microsolv Technology Corporation Supplier No. 9502C40TS
Our Crimp Cap. The Rubber is softer than Natural Red Rubber and provides better sealing after injection. Great for GC. This is our most economical aluminum crimp cap.