Learn More
Microsolv Technology Corporation Caps, Crimp Top, with Red Rubber / PTFE Pre-Slit Septa, fitted in aluminum, Sliver Caps. For use with 11mm Crimp or Snap Top, 12x32mm autosampler vials. 100/PK.
Catalog No. 502270373
Encompass_Preferred
This Pre-Slit Septa is sometimes used in Solid Phase Micro-Extraction (SPME) methods and when 2ml vials are being used.
This item is not returnable. View return policy
Catalog No. 50-227-0373 Supplier Microsolv Technology Corporation Supplier No. 9502C43TS
This Pre-Slit Septa is sometimes used in Solid Phase Micro-Extraction (SPME) methods and when 2ml vials are being used.