Learn More
Microsolv Technology Corporation Caps, Crimp Top, with an Open Hole. Aluminum, Silver Caps. Septa are sold separately and must be inserted manually. For use with 11mm Crimp or Snap Top, 12x32mm autosampler vials. 1000/CS.
Catalog No. 502270361
Encompass_Preferred
Choose Septa for 11mm Caps.
This item is not returnable. View return policy
Catalog No. 50-227-0361 Supplier Microsolv Technology Corporation Supplier No. 9502C00SM
Choose Septa for 11mm Caps.