Learn More
Microsolv Technology Corporation Caps, Crimp Top, with PTFE Septa, fitted in aluminum, Silver Caps. For use with 11mm Crimp or Snap Top, 12x32mm autosampler vials. 100/PK.
Catalog No. 502271253
Encompass_Preferred
This PTFE (only) Septa is for single injections or non-volatile samples as it will not reseal. Buying in case quantities can save money and time.
This item is not returnable. View return policy
Catalog No. 50-227-1253 Supplier Microsolv Technology Corporation Supplier No. 9502C40PS
This PTFE (only) Septa is for single injections or non-volatile samples as it will not reseal. Buying in case quantities can save money and time.