Learn More
Beckman Coulter Microfuge 18 Centrifuge Tubes, 1.5mL, Natural, Bag of 500
Catalog No. NC2380892
Encompass_Preferred
The Beckman Coulter Microfuge 18 Centrifuge is a compact benchtop microcentrifuge designed for quick spin-downs and separations. It has a maximum speed of 13,300 rpm and maximum RCF of 16,060 xg. The rotor accommodates 18 x 1.5/2.0mL microcentrifuge tubes. It features a compact footprint, digital display, and 6 acceleration/deceleration rates for gentle separation. The Microfuge 18 is ideal for DNA/RNA extraction, PCR setup, and other molecular biology applications requiring fast microcentrifuge performance.
Catalog No. NC2380892 Supplier Beckman Coulter Supplier No. 343656
The Beckman Coulter Microfuge 18 Centrifuge is a compact benchtop microcentrifuge designed for quick spin-downs and separations. It has a maximum speed of 13,300 rpm and maximum RCF of 16,060 xg. The rotor accommodates 18 x 1.5/2.0mL microcentrifuge tubes. It features a compact footprint, digital display, and 6 acceleration/deceleration rates for gentle separation. The Microfuge 18 is ideal for DNA/RNA extraction, PCR setup, and other molecular biology applications requiring fast microcentrifuge performance.