Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hach Company Caps For 250ml PE Bottles

Catalog No. 5000257057 Shop All Hach Company Products
Encompass

Caps PTFE lined for 350 ml glass bottles Case of 96

Catalog No. 50-002-57057 Supplier Hach Company Supplier No. 932
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Caps PTFE lined for 350 ml glass bottles Case of 96

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.