Learn More
Microsolv Technology Corporation Caps For 40Ml Epa Vials Sil/Pt
Catalog No. 5001452440
Encompass_Preferred
Caps, Screw Top, with White Silicone Rubber / Beige PTFE Septa, fitted in knurled polypropylene, White Caps. For use with 24-400mm threaded EPA Style vials. ND24 threads. MicroSolv Brand. 100/PK.
Catalog No. 50-014-52440 Supplier Microsolv Technology Corporation Supplier No. 9704010C
Caps, Screw Top, with White Silicone Rubber / Beige PTFE Septa, fitted in knurled polypropylene, White Caps. For use with 24-400mm threaded EPA Style vials. ND24 threads. MicroSolv Brand. 100/PK.