Learn More
Microsolv Technology Corporation Caps For 40Ml Vials Screw Top
Catalog No. 5001452442
Encompass_Preferred
Caps, Screw Top, Solid, with Blue PTFE / EPDM Rubber / Blue PTFE Liner, fitted in knurled polypropylene, White Caps. For use with 24-400mm threaded EPA Style vials. ND24 threads. MicroSolv Brand. 100/PK.
Catalog No. 50-014-52442 Supplier Microsolv Technology Corporation Supplier No. 9704011CS
Caps, Screw Top, Solid, with Blue PTFE / EPDM Rubber / Blue PTFE Liner, fitted in knurled polypropylene, White Caps. For use with 24-400mm threaded EPA Style vials. ND24 threads. MicroSolv Brand. 100/PK.