Learn More
Microsolv Technology Corporation Caps For 40Ml Vials Sil/Pt Blk
Catalog No. 5001452436
Encompass_Preferred
Caps, Screw Top, with White Silicone Rubber / White PTFE Septa, fitted in knurled polypropylene, Black Caps. For use with 24-414mm threaded EPA Style vials. BASIK Brand. 100/PK..
Catalog No. 50-014-52436 Supplier Microsolv Technology Corporation Supplier No. 9704005CK
Caps, Screw Top, with White Silicone Rubber / White PTFE Septa, fitted in knurled polypropylene, Black Caps. For use with 24-414mm threaded EPA Style vials. BASIK Brand. 100/PK..