Learn More
Electron Microscopy Sciences Caps for Syringes, 1000 per pack
SDP
Catalog No. 50949420
Encompass_Preferred
One size fits-all syringe caps that can be used with either a luer slip or luer locking syringe tip. These tips make the syringes suitable for usage as embedding media storage vessels.
Catalog No. 50-949-420 Supplier Electron Microscopy Sciences Supplier No. 7250010
One size fits-all syringe caps that can be used with either a luer slip or luer locking syringe tip. These tips make the syringes suitable for usage as embedding media storage vessels.