Learn More
Microsolv Technology Corporation Caps, Headspace, Crimp Top, with Translucent Silicone Rubber / Natural PTFE, High Purity Septa, fitted in Aluminum, Silver Caps with 10mm center holes. For use with 20mm Crimp Top Headspace Vials. 100/PK.
Catalog No. 502270355
Encompass_Preferred
Silicone Rubber / PTFE Septa with Durometer of 45 Shore A, (hardness) and 3.0mm thick with Ultra Clean PTFE and Ultra Low Bleed Rubber that is made by a proprietary process for ultra high purity, low extractables and temperature durability. This Septa is good for highly sensitive analyses. No color has been added to this Septa. The lower Durometer means that this is a softer Septa and therefore better for easier penetrations by autosampler needles. Wide temperature range from -60⁰ to 200⁰C.
This item is not returnable. View return policy
Catalog No. 50-227-0355 Supplier Microsolv Technology Corporation Supplier No. 95025091S
Silicone Rubber / PTFE Septa with Durometer of 45 Shore A, (hardness) and 3.0mm thick with Ultra Clean PTFE and Ultra Low Bleed Rubber that is made by a proprietary process for ultra high purity, low extractables and temperature durability. This Septa is good for highly sensitive analyses. No color has been added to this Septa. The lower Durometer means that this is a softer Septa and therefore better for easier penetrations by autosampler needles. Wide temperature range from -60⁰ to 200⁰C.