Learn More
Microsolv Technology Corporation Caps, Headspace, Crimp Top, with an Open Hole. Aluminum Silver Caps. Septa are sold separately. For use with 20mm Crimp Top Headspace vials. 100/PK.
Catalog No. 502271196
Encompass_Preferred
Septa are sold separately and must be inserted manually.
This item is not returnable. View return policy
Catalog No. 50-227-1196 Supplier Microsolv Technology Corporation Supplier No. 95025001S
Septa are sold separately and must be inserted manually.