Learn More
Microsolv Technology Corporation Caps, Headspace, Crimp Top, with White Silicone Rubber / Blue PTFE High Purity Septa, fitted in Steel, Magnetic, Gold Caps with 5mm center holes. For use with 20mm Crimp Top Headspace Vials. 100/PK.
Catalog No. 502271211
Encompass_Preferred
Silicone Rubber / PTFE with Durometer of 55 Shore A, (hardness) and 4.5mm thick. Wide temperature range from -60⁰ to 200⁰C. Recommended for CTC™ and other autosamplers.
This item is not returnable. View return policy
Catalog No. 50-227-1211 Supplier Microsolv Technology Corporation Supplier No. 95025233D
Silicone Rubber / PTFE with Durometer of 55 Shore A, (hardness) and 4.5mm thick. Wide temperature range from -60⁰ to 200⁰C. Recommended for CTC™ and other autosamplers.