Learn More
Microsolv Technology Corporation Caps, Headspace, Crimp Top, with White Silicone Rubber / Blue PTFE Septa, fitted in Steel, Magnetic, Gold Caps with 8mm center holes. For use with 20mm Crimp Top Headspace Vials. 100/PK.
Catalog No. 502270275
Encompass_Preferred
Compatible with SPME, Solid Phase Micro-Extraction capable autosamplers including CTC Pal. Silicone Rubber / UltraClean PTFE Septa is 55⁰ Shore A (hardness) and is 1.5mm thick. Use with SPME Vials.
This item is not returnable. View return policy
Catalog No. 50-227-0275 Supplier Microsolv Technology Corporation Supplier No. 95025234D
Compatible with SPME, Solid Phase Micro-Extraction capable autosamplers including CTC Pal. Silicone Rubber / UltraClean PTFE Septa is 55⁰ Shore A (hardness) and is 1.5mm thick. Use with SPME Vials.