Learn More
Microsolv Technology Corporation Caps, Headspace, Crimp Top, with Grey Butyl Rubber / Transparent PTFE Septa, fitted in Steel, Magnetic, Gold Caps with 8mm center holes. For use with 20mm Crimp Top Headspace Vials. 1000/CS.
Catalog No. 502271215
Encompass_Preferred
Butyl Rubber Septa that is laminated with PTFE and is 55⁰ Shore A (hardness) and is 3.0mm thick. Temperature resistant from -40C to 120C.
This item is not returnable. View return policy
Catalog No. 50-227-1215 Supplier Microsolv Technology Corporation Supplier No. 9502560C
Butyl Rubber Septa that is laminated with PTFE and is 55⁰ Shore A (hardness) and is 3.0mm thick. Temperature resistant from -40C to 120C.