Learn More
Microsolv Technology Corporation Caps, Headspace, Crimp Top, with Grey Butyl Rubber / Partially Laminated PTFE Septa, fitted in Aluminum, Silver Caps with 10mm center holes. Pharma-Fix. For use with 20mm Crimp Top Headspace Vials. 100/PK.
Catalog No. 502271202
Encompass_Preferred
Shaped, Butyl Rubber with Durometer of 50 Shore A, (hardness) and 3.0mm thick. Partially laminated with PTFE only on the light grey side of the Septa that faces the sample area. Butyl Rubber facing the glass vial provides a very good seal, due to its elasticity. Combining inertness of PTFE and elasticity of Butyl Rubber. This type of Septa is commonly known as Pharma-Fix™ and is temperature resistant from -40⁰ to 120⁰C.
This item is not returnable. View return policy
Catalog No. 50-227-1202 Supplier Microsolv Technology Corporation Supplier No. 95025031S
Shaped, Butyl Rubber with Durometer of 50 Shore A, (hardness) and 3.0mm thick. Partially laminated with PTFE only on the light grey side of the Septa that faces the sample area. Butyl Rubber facing the glass vial provides a very good seal, due to its elasticity. Combining inertness of PTFE and elasticity of Butyl Rubber. This type of Septa is commonly known as Pharma-Fix™ and is temperature resistant from -40⁰ to 120⁰C.