Learn More
Microsolv Technology Corporation Caps, Headspace, Crimp Top, with Translucent Blue Silicone Rubber / Transparent PTFE Septa, fitted in Steel, Magnetic, Gold Caps with 5mm center holes. For use with 20mm Crimp Top Headspace Vials. 100/PK.
Supplier:  Microsolv Technology Corporation 95025043D
Encompass_Preferred
Silicone Rubber / Ultraclean PTFE with Durometer of 45 Shore A, (hardness) and 3.2mm thick. Completely laminated with PTFE on both sides of the Septa which is good for sensitive analyses. The lower Durometer means that this is a softer Septa and therefore better for easier penetrations by autosampler needles. Wide temperature range from -60⁰ to 200⁰C. Recommended for CTC™ and other autosamplers.
Catalog No. 50-227-1206