Learn More
Microsolv Technology Corporation Caps, Headspace, Crimp Top, with White Silicone Rubber / Blue PTFE SPME Style Septa, fitted in Steel, Magnetic, Gold Caps with 8mm center holes. For use with 20mm Crimp Top Headspace Vials. 100/PK.
Catalog No. 502271212
Encompass_Preferred
Special Shape Septa with thick rubber outer ring (3mm) and thinned rubber center (1.5mm) provides a good seal with most 20mm crimp top Headspace vials. The target area on this Septa is easer for SPME needles to penetrate, minimizing broken or damaged needles. Recommended with CTC Pal and other autosamplers with SPME or Solid Phase Micro-Extraction injectors.
This item is not returnable. View return policy
Catalog No. 50-227-1212 Supplier Microsolv Technology Corporation Supplier No. 95025261D
Special Shape Septa with thick rubber outer ring (3mm) and thinned rubber center (1.5mm) provides a good seal with most 20mm crimp top Headspace vials. The target area on this Septa is easer for SPME needles to penetrate, minimizing broken or damaged needles. Recommended with CTC Pal and other autosamplers with SPME or Solid Phase Micro-Extraction injectors.