Learn More
Microsolv Technology Corporation Caps, Headspace, Crimp Top, with Orange Silicone Rubber / Foil, fitted in Aluminum, Silver Caps with 10mm center holes. For use with 20mm Crimp Top Headspace Vials. 100/PK.
Supplier:  Microsolv Technology Corporation 95025151S
Encompass_Preferred
Silicone Rubber / Aluminum Foil Silver Septa with Durometer of 50 Shore A, (hardness) and 2.5mm thick is good for high temperature analyses. No color has been added to this Septa. Often used with Perkin Elmer Instruments.Wide temperature range from -60⁰ to 220⁰C.
Catalog No. 50-226-9899