Learn More
Microsolv Technology Corporation Caps, Headspace, Screw Top, with White Silicone Rubber / Blue PTFE Septa, fitted in Steel, Magnetic, Silver Caps with 8mm center holes. For use with 18mm Precision Thread Headspace Vials. 1000/CS.
Catalog No. 502270351
Encompass_Preferred
Silicone Rubber / PTFE Septa has durometer of 50⁰ Shore A (hardness) and is 1.5mm thick. Cap has ND18 Screw Threads.
This item is not returnable. View return policy
Catalog No. 50-227-0351 Supplier Microsolv Technology Corporation Supplier No. 9502050SWK
Silicone Rubber / PTFE Septa has durometer of 50⁰ Shore A (hardness) and is 1.5mm thick. Cap has ND18 Screw Threads.