Learn More
Microsolv Technology Corporation Caps, Headspace, Screw Top, with Blue Silicone Rubber / Transparent PTFE Septa, fitted in Steel, Magnetic, Silver Caps with 8mm center holes. For use with 18mm Precision Thread Headspace Vials. 1000/CS.
Catalog No. 502270273
Encompass_Preferred
Silicone Rubber / Ultraclean PTFE Septa has a durometer of 45⁰ Shore A (hardness) and is 1.3mm thick. Cap has ND18 Screw Threads. Recommended with CTC, Agilent, Perkin Elmer TurboMax 16, 40 & 110 and other autosamplers.
This item is not returnable. View return policy
Catalog No. 50-227-0273 Supplier Microsolv Technology Corporation Supplier No. 9502045STK
Silicone Rubber / Ultraclean PTFE Septa has a durometer of 45⁰ Shore A (hardness) and is 1.3mm thick. Cap has ND18 Screw Threads. Recommended with CTC, Agilent, Perkin Elmer TurboMax 16, 40 & 110 and other autosamplers.