Learn More
Microsolv Technology Corporation Caps, Headspace, Screw Top, with Reddish Butyl Rubber / Grey PTFE Septa, fitted in Steel, Magnetic, Silver Caps with 8mm center holes. For use with 18mm Precision Thread Headspace Vials. 1000/CS.
Catalog No. 502270272
Encompass_Preferred
Butyl Rubber / PTFE Septa has a durometer of 55⁰ Shore A (hardness) and is 1.6mm thick. Cap has ND18 Screw Threads. Recommended with Agilent, Perkin Elmer TurboMax 16, 40 & 110 and other autosamplers.
This item is not returnable. View return policy
Catalog No. 50-227-0272 Supplier Microsolv Technology Corporation Supplier No. 9502040TSK
Butyl Rubber / PTFE Septa has a durometer of 55⁰ Shore A (hardness) and is 1.6mm thick. Cap has ND18 Screw Threads. Recommended with Agilent, Perkin Elmer TurboMax 16, 40 & 110 and other autosamplers.