Learn More
Microsolv Technology Corporation Caps Hs Scrw Mag 45S Sil/Pt
Catalog No. 5001451833
Encompass_Preferred
Caps, Headspace, Screw Top, with White Silicone Rubber / Silver Foil Septa, fitted in Steel, Magnetic, Silver Caps with 8mm center holes. For use with 18mm Precision Thread Headspace Vials. MicroSolv Brand. 100/PK.
Catalog No. 50-014-51833 Supplier Microsolv Technology Corporation Supplier No. 9502025ST
Caps, Headspace, Screw Top, with White Silicone Rubber / Silver Foil Septa, fitted in Steel, Magnetic, Silver Caps with 8mm center holes. For use with 18mm Precision Thread Headspace Vials. MicroSolv Brand. 100/PK.