Learn More
Microsolv Technology Corporation Caps Ms Sil/Ptfe Blue
Catalog No. 5001452104
Encompass_Preferred
Caps, Screw Top, with White Silicone Rubber / White PTFE Septa, fitted in knurled, polypropylene, Blue Caps. Recommended for Mass Spec. For use with 9-425mm thread, 12x32mm autosampler vials. Compares to Agilent caps. 100/PK.
Catalog No. 50-014-52104 Supplier Microsolv Technology Corporation Supplier No. 9502S10MB
Caps, Screw Top, with White Silicone Rubber / White PTFE Septa, fitted in knurled, polypropylene, Blue Caps. Recommended for Mass Spec. For use with 9-425mm thread, 12x32mm autosampler vials. Compares to Agilent caps. 100/PK.