Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Caps for Nunclon™ Delta Flasks

Catalog No. 12567253 Shop All Thermo Scientific Products
Change view
Click to view available options
For Use With (Equipment):
175/185/500 cm² Nunclon Delta Flask
25 cm² Nunclon Delta Flask
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
12-567-253 25 cm² Nunclon Delta Flask
12-567-252 175/185/500 cm² Nunclon Delta Flask
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 12-567-253 Supplier Thermo Scientific™ Supplier No. 151152
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Protect your valuable cell culture samples. These caps for Thermo Scientific™ Nunclon™ Delta Flasks are offered in two styles—vent/close, filter—to meet application needs.

Protect your valuable cell culture samples. These caps for Thermo Scientific™ Nunclon™ Delta Flasks are offered in two styles—vent/close (solid) and filter—to meet your application needs.

Vent/close Caps
  • EasY Flask vent/close caps feature a "Y" mark that allows visual verification
  • Vent position easily distinguised even what flasks are stacked in incubators
Filter Caps
  • Filter membrane provides effective barrier against microorganisms
  • Well-defined, hydrophobic filter membrane offers continuous venting with consistent gas exchange and no lot-to-lot variation
  • Chemically robust membrane meets USP class VI standards and is produced without animal-derived substances
Certifications:
  • Sterilized by gamma irradiation and certified to be nonpyrogenic
Ordering Information:
  • Case of 100
TRUSTED_SUSTAINABILITY

Specifications

Closure Color Blue
Description Sterile, HDPE closure
Material HDPE
For Use With (Equipment) 25 cm² Nunclon Delta Flask
No. per Case 100
No. per Pack 1
Product Type Vent/close cap
Sterility Sterile
Class Class VI
Closure Type Solid
Surface Treatment Nunclon Delta
Show More Show Less

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.