Learn More
Microsolv Technology Corporation Caps, Push Top, with Natural Rubber / PTFE Septa, fitted in smooth, polyethylene, Natural Transparent Caps. For use with 8mm, Crimp Top autosampler vials. 100/PK.
Supplier:  Microsolv Technology Corporation 9508C10CC
Encompass_Preferred
This cap is similar to a Snap Cap but does not need the Snap Ring on a vial to create a tight seal.
Catalog No. 50-227-0284
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.