Learn More
Microsolv Technology Corporation Caps, Push Top, Solid, Translucent Polyethylene. No liner required with ND22 Push Top, Econo-Storage Vials. 100/PK.
Catalog No. 502270283
Encompass_Preferred
Transparent Polyethylene Cap snaps on to an Econo Push Top Storage Vials with ND22 beveled Tops. Liners are not required because the Cap is solid and seals with the Snap Top device of the vial. Cap has 23.5x5.5mm outer dimensions.
This item is not returnable. View return policy
Catalog No. 50-227-0283 Supplier Microsolv Technology Corporation Supplier No. 97010SC22
Transparent Polyethylene Cap snaps on to an Econo Push Top Storage Vials with ND22 beveled Tops. Liners are not required because the Cap is solid and seals with the Snap Top device of the vial. Cap has 23.5x5.5mm outer dimensions.