Learn More
Microsolv Technology Corporation Caps, Push Top, with thinned penetration point, inserted in smooth, polyethylene, Blue Caps. For use with 11mm Crimp or Snap Top, 12x32mm autosampler vials. 100/PK.
Catalog No. 502271262
Encompass_Preferred
This cap is useful for non volatile solvents or when evaporation is not considered a problem.
This item is not returnable. View return policy
Catalog No. 50-227-1262 Supplier Microsolv Technology Corporation Supplier No. 9502C90CB
This cap is useful for non volatile solvents or when evaporation is not considered a problem.