Learn More
Thermo Scientific™ Caps for Samco™ Histology Bio-Tite™ Nonsterile Specimen Containers

Description
Prevent bio-hazardous spills with these Caps for Thermo Scientific™ Samco™ Histology Bio-Tite™ Specimen Containers.
- Bio-Tite caps create leak-resistant seal for superior safety and handling
- Dual-threaded caps create improved sealing
Ordering Information:
Bulk packed, one bag per case
Specifications
Specifications
| Product Type | Specimen Container |
| Closure Included | Yes |
| Shape | Cylindrical |
| Sterility | Non-sterile |
| Closure Color | White |
| Closure Size | 53 mm |
| Closure Type | Bio-Tite Lid |
| Description | Caps, 53mm; White |
| Diameter (Metric) | 53 mm |
| No. per Case | 1500/Cs. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.