Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Caps for Samco™ Histology Bio-Tite™ Nonsterile Specimen Containers
GREENER_CHOICE

Catalog No. p-6013210
Change view
Click to view available options
Closure Size:
48 mm
53 mm
Closure Color:
White
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Closure Size Closure Color
13-711-93 53 mm White
13-711-91 48 mm White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 13-711-93 Supplier Thermo Scientific™ Supplier No. 5A 0052
Only null left
Add to Cart
Add to Cart

Prevent bio-hazardous spills with these Caps for Thermo Scientific™ Samco™ Histology Bio-Tite™ Specimen Containers.

Prevent bio-hazardous spills with these Caps for Thermo Scientific™ Samco™ Histology Bio-Tite™ Specimen Containers.

  • Bio-Tite caps create leak-resistant seal for superior safety and handling
  • Dual-threaded caps create improved sealing

Ordering Information:

Bulk packed, one bag per case

TRUSTED_SUSTAINABILITY

Specifications

Product Type Specimen Container
Closure Included Yes
Shape Cylindrical
Sterility Non-sterile
Closure Color White
Closure Size 53 mm
Closure Type Bio-Tite Lid
Description Caps, 53mm; White
Diameter (Metric) 53 mm
No. per Case 1500/Cs.
No. per Pack 1500/Pk.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.