Learn More
Microsolv Technology Corporation Caps, Screw Top, with 1.35mm thick, Silicone Rubber / PTFE Septa fitted in "knurled", polypropylene, Light Blue Caps. For use with 10-425mm thread, 12x32mm wide mouth autosampler vials. 100/PK
Catalog No. 502270430
Encompass_Preferred
Caps, Screw Top, with 1.35mm thick, Silicone Rubber / PTFE Septa fitted in "knurled", polypropylene, Light Blue Caps. For use with 10-425mm thread, 12x32mm wide mouth autosampler vials. 100/PK
This item is not returnable. View return policy
Catalog No. 50-227-0430 Supplier Microsolv Technology Corporation Supplier No. 9503SE11CLB
Caps, Screw Top, with 1.35mm thick, Silicone Rubber / PTFE Septa fitted in "knurled", polypropylene, Light Blue Caps. For use with 10-425mm thread, 12x32mm wide mouth autosampler vials. 100/PK