Learn More
Microsolv Technology Corporation Caps, Screw Top, with 1.3mm thick Silicone Rubber / PTFE Pre-Slit Septa, fitted in "knurled", polypropylene, Black Caps with 5.5mm center holes. For use with 8-425mm thread, 12x32mm autosampler vials. 100/PK.
Catalog No. 502271277
Encompass_Preferred
Pre-Slit for easy penetration by autosampler needles.
This item is not returnable. View return policy
Catalog No. 50-227-1277 Supplier Microsolv Technology Corporation Supplier No. 9582S30CK
Pre-Slit for easy penetration by autosampler needles.