Learn More
Microsolv Technology Corporation Caps Screw 10-425 Sil/Ptfe Sh
Catalog No. 5001452208
Encompass_Preferred
Caps, Screw Top, with Silicone Rubber / PTFE Septa fitted in knurled, polypropylene, Black Caps. For use with 10-425mm thread, 12x32mm wide mouth autosampler vials. Compare to Shimadzu Caps. BASIK Brand. 100/PK
Catalog No. 50-014-52208 Supplier Microsolv Technology Corporation Supplier No. 9503SGR
Caps, Screw Top, with Silicone Rubber / PTFE Septa fitted in knurled, polypropylene, Black Caps. For use with 10-425mm thread, 12x32mm wide mouth autosampler vials. Compare to Shimadzu Caps. BASIK Brand. 100/PK