Learn More
Microsolv Technology Corporation Caps Screw 8-425 Sil/Ptfe Bla
Catalog No. 5001452401
Encompass_Preferred
Caps, Screw Top, with Silicone Rubber / PTFE Septa, fitted in knurled, polypropylene, Black Caps. For use with 8-425mm thread, 12x32mm autosampler vials. BASIK Brand. 100/PK.
Catalog No. 50-014-52401 Supplier Microsolv Technology Corporation Supplier No. 9582S05B
Caps, Screw Top, with Silicone Rubber / PTFE Septa, fitted in knurled, polypropylene, Black Caps. For use with 8-425mm thread, 12x32mm autosampler vials. BASIK Brand. 100/PK.