Learn More
Microsolv Technology Corporation Caps Screw 9-425 Pe Single Inj
Catalog No. 5001452086
Encompass_Preferred
Caps, Screw Top, Solid, Single injection use with a thinned penetration point. No Septa required. LCMS compatible, knurled polyethylene, Natural color Caps. For use with with 9-425mm thread, 12x32mm autosampler vials. AQ Brand. 100/PK.
Catalog No. 50-014-52086 Supplier Microsolv Technology Corporation Supplier No. 9502S09PE
Caps, Screw Top, Solid, Single injection use with a thinned penetration point. No Septa required. LCMS compatible, knurled polyethylene, Natural color Caps. For use with with 9-425mm thread, 12x32mm autosampler vials. AQ Brand. 100/PK.