Learn More
Microsolv Technology Corporation Caps Screw Black Bondedcaps
Catalog No. 502764
Encompass_Preferred
Caps, Screw Top, with Silicone Rubber / PTFE, low durometer Septa, bonded in knurled, polypropylene, Black Caps. For use with 9-425mm thread, 12x32mm autosampler vials. MicroSolv Brand. 100/PK.
Catalog No. 50-276-4 Supplier Microsolv Technology Corporation Supplier No. 9502S10CBLK
Caps, Screw Top, with Silicone Rubber / PTFE, low durometer Septa, bonded in knurled, polypropylene, Black Caps. For use with 9-425mm thread, 12x32mm autosampler vials. MicroSolv Brand. 100/PK.