Learn More
Microsolv Technology Corporation Caps Screw Blue P/S/P Septaca
Catalog No. 502768
Encompass_Preferred
Caps, Screw Top, with Red PTFE / White Silicone Rubber / Red PTFE Septa, fitted in knurled, polypropylene, Blue Caps. For use with 9-425mm thread, 12x32mm autosampler vials. MicroSolv Brand. 1000/CS
Catalog No. 50-276-8 Supplier Microsolv Technology Corporation Supplier No. 9502S20CBM
Caps, Screw Top, with Red PTFE / White Silicone Rubber / Red PTFE Septa, fitted in knurled, polypropylene, Blue Caps. For use with 9-425mm thread, 12x32mm autosampler vials. MicroSolv Brand. 1000/CS