Learn More
Microsolv Technology Corporation Caps,Screw Cap,Ptfe Septa C/Pk
Catalog No. NC1344221
Encompass_Preferred
Caps, Screw Top, with Soft-Guard, White Silicone Rubber / Red PTFE Septa, bonded in knurled, polypropylene, Blue Caps. For use with 9-425mm thread, 12x32mm autosampler vials. Soft-Guard Brand. 100/PK
Catalog No. NC1344221 Supplier Microsolv Technology Corporation Supplier No. 9502SE10CB
Caps, Screw Top, with Soft-Guard, White Silicone Rubber / Red PTFE Septa, bonded in knurled, polypropylene, Blue Caps. For use with 9-425mm thread, 12x32mm autosampler vials. Soft-Guard Brand. 100/PK