Learn More
Microsolv Technology Corporation Caps Screw Lc-Clean Sil/Ptfe
Catalog No. 5001452081
Encompass_Preferred
Caps, Screw Top, with Clear Silicone Rubber / PTFE, ultra-pure, LC-MS compatible LC Clean Septa, bonded in knurled, polypropylene, Blue Caps. For use with 9-425mm thread, 12x32mm autosampler vials. MicroSolv Brand. 100/PK.
Catalog No. 50-014-52081 Supplier Microsolv Technology Corporation Supplier No. 9502S06LCC
Caps, Screw Top, with Clear Silicone Rubber / PTFE, ultra-pure, LC-MS compatible LC Clean Septa, bonded in knurled, polypropylene, Blue Caps. For use with 9-425mm thread, 12x32mm autosampler vials. MicroSolv Brand. 100/PK.