Learn More
Microsolv Technology Corporation Caps Screw Pp 15-425 9Mm Open
Catalog No. 5001452420
Encompass_Preferred
Caps, Screw Top, with White Silicone Rubber / Red PTFE Septa / Liner, fitted in knurled polypropylene, Black Caps. For use with ND15 thread, storage Vials or Bottles. MicroSolv Brand. 100/PK.
Catalog No. 50-014-52420 Supplier Microsolv Technology Corporation Supplier No. 9701510K
Caps, Screw Top, with White Silicone Rubber / Red PTFE Septa / Liner, fitted in knurled polypropylene, Black Caps. For use with ND15 thread, storage Vials or Bottles. MicroSolv Brand. 100/PK.