Learn More
Microsolv Technology Corporation Caps, Screw Top, with Red PTFE / White Silicone Rubber / Red PTFE Septa, fitted in "knurled", polypropylene, Green Caps. For use with 9-425mm thread, 12x32mm autosampler vials. 1000/CS
Catalog No. 502271298
Encompass_Preferred
This septa was designed for specific, older instruments such as Agilent 1050 and to provide extra cleaning when "wiping" a needle. Also provides a small degree of better resealing after injection compared to Silicone Rubber / PTFE Septa.
This item is not returnable. View return policy
Catalog No. 50-227-1298 Supplier Microsolv Technology Corporation Supplier No. 9502S20CGM
This septa was designed for specific, older instruments such as Agilent 1050 and to provide extra cleaning when "wiping" a needle. Also provides a small degree of better resealing after injection compared to Silicone Rubber / PTFE Septa.