Learn More
Microsolv Technology Corporation Caps, Screw Top, Solid PTFE with thinned penetration area. "Knurled" Translucent Caps. For use with 9-425mm thread, 12x32mm autosampler vials. 100/PK.
Catalog No. 502271267
Encompass_Preferred
This low-extractables, PTFE Cap is chosen when Polypropylene and Silicone Rubber cannot be used in your analysis.
This item is not returnable. View return policy
Catalog No. 50-227-1267 Supplier Microsolv Technology Corporation Supplier No. 9502S50N
This low-extractables, PTFE Cap is chosen when Polypropylene and Silicone Rubber cannot be used in your analysis.