Learn More
Microsolv Technology Corporation Caps, Screw Top, with Natural Rubber / TFE Septa, fitted in "knurled", polypropylene, Blue Caps. For use with 9-425mm thread, 12x32mm autosampler vials. 1000/CS.
Catalog No. 502271284
Encompass_Preferred
Natural Rubber does not contain siloxanes like Silicone Rubber. TFE is similar to PTFE when comparing inertness.
This item is not returnable. View return policy
Catalog No. 50-227-1284 Supplier Microsolv Technology Corporation Supplier No. 9502S80CBM
Natural Rubber does not contain siloxanes like Silicone Rubber. TFE is similar to PTFE when comparing inertness.