Learn More
Microsolv Technology Corporation Caps, Screw Top, with Beige Silicone Rubber / White PTFE ultra-pure Septa, fitted in "knurled", polypropylene, Blue Caps with 6mm holes. For use with 9-425 thread, 12x32mm autosampler vials. 100/PK.
Catalog No. 502270286
Encompass_Preferred
Larger, 6mm hole in the cap provides a larger target area for autosamplers that might have difficulty and may strike the cap edge of smaller hole caps, during injection.
This item is not returnable. View return policy
Catalog No. 50-227-0286 Supplier Microsolv Technology Corporation Supplier No. 9502S36B
Larger, 6mm hole in the cap provides a larger target area for autosamplers that might have difficulty and may strike the cap edge of smaller hole caps, during injection.