Learn More
Microsolv Technology Corporation Caps, Screw Top, with White Silicone Rubber / White PTFE Pre-Slit Septa, fitted in "knurled", polypropylene, Blue Caps. For use with 9-425mm thread, 12x32mm autosampler vials. 100/PK
Catalog No. 502270297
Encompass_Preferred
Pre-slit for easy penetration by autosampler needles. Recommended for Waters Alliance & Acquity instruments. Ultra-pure Silicone Rubber and "Cast" PTFE for excellent results in LCMS/MS compares to LCMS Certified brands.
This item is not returnable. View return policy
Catalog No. 50-227-0297 Supplier Microsolv Technology Corporation Supplier No. 9502S30MB
Pre-slit for easy penetration by autosampler needles. Recommended for Waters Alliance & Acquity instruments. Ultra-pure Silicone Rubber and "Cast" PTFE for excellent results in LCMS/MS compares to LCMS Certified brands.