Learn More
Microsolv Technology Corporation Caps, Screw Top, with Natural Silicone Rubber / Natural PTFE Septa, fitted in knurled polypropylene, White Caps. For use with 24-414mm threaded EPA Style vials. ND24 threads. 100/PK.
Catalog No. 502271285
Encompass_Preferred
Septa is 3.18mm (0.125") thick and can be used as an autosampler vial cap or for long term storage with any 30, 40 or 60 EPA Style vial that has 40-414 threads. Cap has a 12.5mm top hole.
This item is not returnable. View return policy
Catalog No. 50-227-1285 Supplier Microsolv Technology Corporation Supplier No. 97041SC24
Septa is 3.18mm (0.125") thick and can be used as an autosampler vial cap or for long term storage with any 30, 40 or 60 EPA Style vial that has 40-414 threads. Cap has a 12.5mm top hole.